3-[([1,1'-Biphenyl]-4-yloxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050127-500MG 050127-2.5G
101890-408EA 364.62 USD
101890-408 101890-410
3-[([1,1'-Biphenyl]-4-yloxy)methyl]pyrrolidine hydrochloride
3-[([1,1'-Biphenyl]-4-yloxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀ClNO
MDL Number: MFCD13560529
CAS Number: 1220037-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR