2-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050126-500MG 050126-2.5G
101890-404EA 186.15 USD
101890-404 101890-406
2-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride
2-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD13560528
CAS Number: 1219963-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR