3-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050125-500MG 050125-2.5G
101890-400EA 364.62 USD
101890-400 101890-402
3-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride
3-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD13560527
CAS Number: 1220032-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR