4-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050124-500MG 050124-2.5G
101890-396EA 364.62 USD
101890-396 101890-398
4-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride
4-[2-([1,1'-Biphenyl]-4-yloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₄ClNO
MDL Number: MFCD13560526
CAS Number: 1219971-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR