3-[(4-Propoxyphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050123-500MG
050123-2.5G
101890-392EA
364.62
USD
101890-392
101890-394
3-[(4-Propoxyphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Propoxyphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO₂ |
MDL Number:
MFCD13560525 CAS Number: 1220032-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...