2-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050114-500MG 050114-2.5G
101890-356EA 364.62 USD
101890-356 101890-358
2-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride
2-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₆ClNO₂
MDL Number: MFCD13560516
CAS Number: 158550-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR