3-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050113-500MG 050113-2.5G
101890-352EA 364.62 USD
101890-352 101890-354
3-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride
3-{2-[4-(Benzyloxy)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₆ClNO₂
MDL Number: MFCD13560515
CAS Number: 1219961-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR