N-[4-(3-Pyrrolidinylmethoxy)phenyl]acetamide hydrochloride
Supplier: Matrix Scientific
050111-500MG
050111-2.5G
101890-344EA
364.62
USD
101890-344
101890-346
N-[4-(3-Pyrrolidinylmethoxy)phenyl]acetamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂ |
MDL Number:
MFCD13560513 CAS Number: 1219961-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...