N-{4-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050110-500MG 050110-2.5G
101890-340EA 364.62 USD
101890-340 101890-342
N-{4-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{4-[2-(2-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560512
CAS Number: 879662-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR