N-{4-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050109-500MG 050109-2.5G
101890-336EA 364.62 USD
101890-336 101890-338
N-{4-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{4-[2-(3-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560511
CAS Number: 1220016-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR