N-{4-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Supplier: Matrix Scientific

050108-500MG 050108-2.5G
101890-332EA 364.62 USD
101890-332 101890-334
N-{4-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride
N-{4-[2-(4-Piperidinyl)ethoxy]phenyl}acetamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂O₂
MDL Number: MFCD13560510
CAS Number: 1219971-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR