2-[2-(4-Iodophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050106-500MG 050106-2.5G
101890-324EA 364.62 USD
101890-324 101890-326
2-[2-(4-Iodophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Iodophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClINO
MDL Number: MFCD13560508
CAS Number: 1220020-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR