3-[(4-Chloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050103-500MG 050103-2.5G
101890-312EA 364.62 USD
101890-312 101890-314
3-[(4-Chloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Chloro-3,5-dimethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560505
CAS Number: 1219971-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR