4-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050100-500MG 050100-2.5G
101890-300EA 364.62 USD
101890-300 101890-302
4-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560502
CAS Number: 1220032-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR