4-Chloro-3-methylphenyl-3-pyrrolidinylmethylether hydrochloride

Supplier: Matrix Scientific

050099-500MG 050099-2.5G
101890-296EA 364.62 USD
101890-296 101890-298
4-Chloro-3-methylphenyl-3-pyrrolidinylmethylether hydrochloride
4-Chloro-3-methylphenyl-3-pyrrolidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MW: 262.18 g/mol
MDL Number: MFCD13560501
CAS Number: 1220032-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR