2-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050098-500MG 050098-2.5G
101890-292EA 364.62 USD
101890-292 101890-294
2-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560500
CAS Number: 1220019-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR