3-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050097-500MG 050097-2.5G
101890-288EA 364.62 USD
101890-288 101890-290
3-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560499
CAS Number: 1219982-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR