4-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050096-500MG 050096-2.5G
101890-284EA 364.62 USD
101890-284 101890-286
4-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Chloro-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560498
CAS Number: 1219981-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR