3-[(2-Bromo-4-chlorophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050095-500MG 050095-2.5G
101890-280EA 364.62 USD
101890-280 101890-282
3-[(2-Bromo-4-chlorophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Bromo-4-chlorophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrCl₂NO
MDL Number: MFCD13560497
CAS Number: 1219971-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR