2-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050094-500MG 050094-2.5G
101890-276EA 364.62 USD
101890-276 101890-278
2-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrCl₂NO
MDL Number: MFCD13560496
CAS Number: 1219982-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR