3-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050093-500MG 050093-2.5G
101890-272EA 364.62 USD
101890-272 101890-274
3-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromo-4-chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrCl₂NO
MDL Number: MFCD13560495
CAS Number: 1220032-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR