2-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050090-500MG
050090-2.5G
101890-260EA
364.62
USD
101890-260
101890-262
2-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉Cl₂NO |
MDL Number:
MFCD13560492 CAS Number: 72834-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...