3-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050089-500MG 050089-2.5G
101890-256EA 364.62 USD
101890-256 101890-258
3-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chlorophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560491
CAS Number: 1220019-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR