3-[(4-Bromophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050087-500MG 050087-2.5G
101890-248EA 364.62 USD
101890-248 101890-250
3-[(4-Bromophenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO
MDL Number: MFCD13560489
CAS Number: 1219982-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR