2-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050086-500MG 050086-2.5G
101890-244EA 364.62 USD
101890-244 101890-246
2-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560488
CAS Number: 1220032-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR