3-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050085-500MG
050085-2.5G
101890-240EA
364.62
USD
101890-240
101890-242
3-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉BrClNO |
MDL Number:
MFCD13560487 CAS Number: 1219982-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...