4-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050084-500MG 050084-2.5G
101890-236EA 364.62 USD
101890-236 101890-238
4-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD13560486
CAS Number: 1220019-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR