3-[(2-Isopropyl-5-methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050079-500MG 050079-2.5G
101890-216EA 364.62 USD
101890-216 101890-218
3-[(2-Isopropyl-5-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Isopropyl-5-methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560481
CAS Number: 1220019-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR