2-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050078-500MG 050078-2.5G
101890-212EA 364.62 USD
101890-212 101890-214
2-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560480
CAS Number: 1219949-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR