3-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050077-500MG 050077-2.5G
101890-208EA 364.62 USD
101890-208 101890-210
3-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560479
CAS Number: 1219982-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR