4-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050076-500MG 050076-2.5G
101890-204EA 364.62 USD
101890-204 101890-206
4-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Isopropyl-5-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560478
CAS Number: 1219981-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR