3-[(3,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
050075-500MG
050075-2.5G
101890-200EA
364.62
USD
101890-200
101890-202
3-[(3,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(3,5-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClNO |
MDL Number:
MFCD13560477 CAS Number: 1220019-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...