2-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050074-500MG 050074-2.5G
101890-196EA 364.62 USD
101890-196 101890-198
2-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560476
CAS Number: 1219961-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR