3-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050073-500MG
050073-2.5G
101890-192EA
364.62
USD
101890-192
101890-194
3-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560475 CAS Number: 1219980-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...