4-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050072-500MG
050072-2.5G
101890-188EA
364.62
USD
101890-188
101890-190
4-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(3,5-Dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560474 CAS Number: 1219982-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...