3-[(3,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050071-500MG 050071-2.5G
101890-184EA 364.62 USD
101890-184 101890-186
3-[(3,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(3,4-Dimethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13560473
CAS Number: 1219980-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR