2-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050070-500MG
050070-2.5G
101890-180EA
364.62
USD
101890-180
101890-182
2-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₄ClNO |
MDL Number:
MFCD13560472 CAS Number: 1219949-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...