3-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050069-500MG 050069-2.5G
101890-176EA 364.62 USD
101890-176 101890-178
3-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(3,4-Dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560471
CAS Number: 1220032-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR