3-[(3-Isopropylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050067-500MG 050067-2.5G
101890-168EA 364.62 USD
101890-168 101890-170
3-[(3-Isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(3-Isopropylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13560469
CAS Number: 1219949-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR