2-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050066-500MG 050066-2.5G
101890-164EA 364.62 USD
101890-164 101890-166
2-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560468
CAS Number: 1219982-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR