3-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050065-500MG
050065-2.5G
101890-160EA
166.15
USD
101890-160
101890-162
3-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride
3-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13560467 CAS Number: 1219982-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...