4-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050064-500MG 050064-2.5G
101890-156EA 364.62 USD
101890-156 101890-158
4-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride
4-[2-(3-Isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560466
CAS Number: 1220031-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR