3-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050063-500MG 050063-2.5G
101890-152EA 364.62 USD
101890-152 101890-154
3-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride
3-(tert-Butyl)phenyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560465
CAS Number: 1219982-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR