2-{2-[3-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050062-500MG 050062-2.5G
101890-148EA 364.62 USD
101890-148 101890-150
2-{2-[3-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[3-(tert-Butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560464
CAS Number: 1219980-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR