3-{[3-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050059-500MG 050059-2.5G
101890-136EA 364.62 USD
101890-136 101890-138
3-{[3-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[3-(Trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃NO
MDL Number: MFCD13560461
CAS Number: 1219982-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR