2-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050058-500MG 050058-2.5G
101890-132EA 364.62 USD
101890-132 101890-134
2-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560460
CAS Number: 72834-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR