3-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050057-500MG 050057-2.5G
101890-128EA 364.62 USD
101890-128 101890-130
3-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560459
CAS Number: 1219949-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR