4-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050056-500MG 050056-2.5G
101890-124EA 364.62 USD
101890-124 101890-126
4-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[3-(Trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClF₃NO
MDL Number: MFCD13560458
CAS Number: 1219968-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR