Ethyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

050052-500MG 050052-2.5G
101890-108EA 364.62 USD
101890-108 101890-110
Ethyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride
Ethyl-3-[2-(4-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO₃
MDL Number: MFCD13560454
CAS Number: 1220031-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR