Methyl-3-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Supplier: Matrix Scientific

050050-500MG 050050-2.5G
101890-100EA 364.62 USD
101890-100 101890-102
Methyl-3-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Methyl-3-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₃
MDL Number: MFCD13560452
CAS Number: 1219981-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR